Stampa…

4-Methylumbelliferyl alpha-D-mannopyranoside ≥97.0% (by HPLC)

Fornitore: TCI

M3023-25MG M3023-100MG
TCIAM3023-25MGEA 61 EUR
TCIAM3023-25MG TCIAM3023-100MG
4-Methylumbelliferyl alpha-D-mannopyranoside ≥97.0% (by HPLC)
4-Methylumbelliferyl alpha-D-mannopyranoside
Formula: C₁₆H₁₈O₈
Peso molecolare: 338.31 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00067662
CAS Number: 28541-83-5
EINECS: 249-073-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=703af9d0d85d6979aee7a01d4bc9d11b0f60a615&vid=703af9d0d85d6979aee7a01d4bc9d11b0f60a615&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 214.0 to 219.0 deg-C
Specific rotation [a]20/D +157 to +167 deg (C=0.35, H2O)
Specific rotation (value) 157 to 167 deg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR