5-Methylpyrazin-2-amine ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Amino-5-methylpyrazine
M3006-1G
M3006-5G
TCIAM3006-1GEA
76
EUR
TCIAM3006-1G
TCIAM3006-5G
5-Methylpyrazin-2-amine ≥99.0% (by GC)
5-Methylpyrazin-2-amine
Formula:
C5H7N3 Peso molecolare: 109,13 g/mol |
MDL Number:
MFCD08437664 CAS Number: 5521-58-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...