Stampa…

5-Methyl-3-phenylisoxazole-4-carbonyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

M2996-1G M2996-5G
TCIAM2996-1GEA 30 EUR
TCIAM2996-1G TCIAM2996-5G
5-Methyl-3-phenylisoxazole-4-carbonyl chloride ≥98.0% (by GC, titration analysis)
5-Methyl-3-phenylisoxazole-4-carbonyl chloride
Formula: C₁₁H₈ClNO₂
Peso molecolare: 221,64 g/mol
Punto di ebollizione: 170°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00052205
CAS Number: 16883-16-2
EINECS: 240-915-4
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR