Stampa…

3-Anisoyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

M2992-5G M2992-25G
TCIAM2992-5GEA 25 EUR
TCIAM2992-5G TCIAM2992-25G
3-Anisoyl chloride ≥98.0% (by GC, titration analysis)
3-Anisoyl chloride
Formula: C₈H₇ClO₂
Punto di ebollizione: 125°C
Flash Pt: 92°C
MDL Number: MFCD00000673
CAS Number: 1711-05-3
EINECS: 216-975-2
UN: 1729
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84bfa14a04a86e378a06e9db7c70a850612b090d&vid=84bfa14a04a86e378a06e9db7c70a850612b090d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR