Stampa…

N-[2-(Methacryloyloxy)ethyl]-N,N-dimethylbutan-1-aminium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Butyl[2-(methacryloyloxy)ethyl](dimethyl)ammonium bis(trifluoromethanesulphonyl)imide

M2987-1G M2987-5G
TCIAM2987-1GEA 62 EUR
TCIAM2987-1G TCIAM2987-5G
N-[2-(Methacryloyloxy)ethyl]-N,N-dimethylbutan-1-aminium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, total nitrogen)
N-[2-(Methacryloyloxy)ethyl]-N,N-dimethylbutan-1-aminium bis(trifluoromethanesulphonyl)imide
Formula: C₁₄H₂₄F₆N₂O₆S₂
Peso molecolare: 494,46 g/mol
CAS Number: 1616483-28-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR