Methyl-2,3-diaminobenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2,3-diaminobenzoate
M2979-1G
M2979-5G
TCIAM2979-1GEA
54
EUR
TCIAM2979-1G
TCIAM2979-5G
Methyl-2,3-diaminobenzoate ≥98.0% (by GC)
Methyl-2,3-diaminobenzoate
Formula:
C₈H₁₀N₂O₂ Peso molecolare: 166,1800 g/mol |
MDL Number:
MFCD04038589 CAS Number: 107582-20-7 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...