Methyl-3-hydroxypicolinate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
M2973-1G
M2973-5G
TCIAM2973-1GEA
67
EUR
TCIAM2973-1G
TCIAM2973-5G
Methyl-3-hydroxypicolinate ≥98.0% (by GC)
Methyl-3-hydroxypicolinate
Formula:
C₇H₇NO₃ |
MDL Number:
MFCD00661311 CAS Number: 62733-99-7 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...