Stampa…

Mupirocin ≥98.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: 9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid

M2955-25MG M2955-100MG
TCIAM2955-25MGEA 70 EUR
TCIAM2955-25MG TCIAM2955-100MG
Mupirocin ≥98.0% (by titrimetric analysis)
Mupirocin
Formula: C₂₆H₄₄O₉
Punto di fusione: 78°C
MDL Number: MFCD01711620
CAS Number: 12650-69-0
Merck Index: 14,06302
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94435663da246822abdc5c2c267bfec5e25c558d&vid=94435663da246822abdc5c2c267bfec5e25c558d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -18.0 to -21.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR