Stampa…

Methyl 2-Nitrophenyl sulphone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-(Methylsulphonyl)-2-nitrobenzene

M2927-1G M2927-5G
TCIAM2927-1GEA 101 EUR
TCIAM2927-1G TCIAM2927-5G
Methyl 2-Nitrophenyl sulphone ≥98.0% (by GC)
Methyl 2-Nitrophenyl sulphone
Formula: C₇H₇NO₄S
Peso molecolare: 201,20 g/mol
MDL Number: MFCD12828669
CAS Number: 2976-34-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR