Stampa…

Methyl 3-Hydroxyphenylacetate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 2-(3-hydroxyphenyl)acetate, 3-Hydroxyphenylacetic acid methyl ester

M2925-1G M2925-5G
TCIAM2925-1GEA 40 EUR
TCIAM2925-1G TCIAM2925-5G
Methyl 3-Hydroxyphenylacetate ≥98.0% (by GC)
Methyl 3-Hydroxyphenylacetate
Formula: C₉H₁₀O₃
Peso molecolare: 166,18 g/mol
Punto di ebollizione: 146°C
MDL Number: MFCD00017660
CAS Number: 42058-59-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=528aca908beea52b6bd633e2f340aeed088c14f2&vid=528aca908beea52b6bd633e2f340aeed088c14f2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR