Stampa…

4-Methyl-3-nitrobenzyl alcohol ≥96.0% (by GC)

Fornitore: TCI

Synonyms: (4-Methyl-3-nitrophenyl)methanol

M2916-5G M2916-25G
TCIAM2916-5GEA 57 EUR
TCIAM2916-5G TCIAM2916-25G
4-Methyl-3-nitrobenzyl alcohol ≥96.0% (by GC)
4-Methyl-3-nitrobenzyl alcohol
Formula: C₈H₉NO₃
Punto di ebollizione: 195°C
MDL Number: MFCD00007177
CAS Number: 40870-59-5
EINECS: 250-120-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6dd8e7021dac15f22cf3384fffce5e2474f0ece&vid=b6dd8e7021dac15f22cf3384fffce5e2474f0ece&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR