Stampa…

(2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Mesityl(o-tolyl)iodonium triflate, (2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulphonate

M2907-1G M2907-5G
TCIAM2907-1GEA 59 EUR
TCIAM2907-1G TCIAM2907-5G
(2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
(2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate
Formula: C₁₇H₁₈F₃IO₃S
Peso molecolare: 486,29 g/mol
MDL Number: MFCD20264884
CAS Number: 210823-54-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52862804cfdfbeb572ecaf631f391357c673ca6c&vid=52862804cfdfbeb572ecaf631f391357c673ca6c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR