Methyl-4-methoxy-2-methylbenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl 4-methoxy-2-methylbenzoate
M2889-1G
M2889-5G
TCIAM2889-1GEA
52
EUR
TCIAM2889-1G
TCIAM2889-5G
Methyl-4-methoxy-2-methylbenzoate ≥98.0% (by GC)
Methyl-4-methoxy-2-methylbenzoate
Formula:
C₁₀H₁₂O₃ Peso molecolare: 180,21 g/mol Punto di ebollizione: 98°C |
MDL Number:
MFCD06203918 CAS Number: 35598-05-1 |
Risultati del test di specificazione
Appearance | White to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Acid Value | max. 3.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...