Stampa…

Methyl-1-benzylpyrrolidine-3-carboxylate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Methyl 1-benzylpyrrolidine-3-carboxylate

M2866-1G M2866-5G
TCIAM2866-1GEA 40 EUR
TCIAM2866-1G TCIAM2866-5G
Methyl-1-benzylpyrrolidine-3-carboxylate ≥98.0% (by GC, titration analysis)
Methyl-1-benzylpyrrolidine-3-carboxylate
Formula: C₁₃H₁₇NO₂
Peso molecolare: 219,28 g/mol
Punto di ebollizione: 105°C
MDL Number: MFCD04038678
CAS Number: 17012-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfc7d91d3d22bb00bb898ea413f885235021b0e4&vid=cfc7d91d3d22bb00bb898ea413f885235021b0e4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR