Stampa…

1-Methyl-2-(methylsulphonyl)benzimidazole ≥98.0% (by GC)

Fornitore: TCI EUROPE

M2860-1G M2860-5G
TCIAM2860-1GEA 108 EUR
TCIAM2860-1G TCIAM2860-5G
1-Methyl-2-(methylsulphonyl)benzimidazole ≥98.0% (by GC)
1-Methyl-2-(methylsulphonyl)benzimidazole
Formula: C₉H₁₀N₂O₂S
Peso molecolare: 210,25 g/mol
CAS Number: 61078-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4042dc33e1e1762e3ab0f6f94f0c1e908439926&vid=d4042dc33e1e1762e3ab0f6f94f0c1e908439926&prid=web&clid=SAPDAM

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR