Stampa…

4-Methoxycarbonyl-2-nitrophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Methoxycarbonyl)-2-nitrophenylboronic acid, 4-(Methoxycarbonyl)-2-nitrobenzeneboronic acid, 4-Methoxycarbonyl-2-nitrobenzeneboronic acid

M2854-1G M2854-5G
TCIAM2854-1GEA 27 EUR
TCIAM2854-1G TCIAM2854-5G
4-Methoxycarbonyl-2-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Methoxycarbonyl-2-nitrophenylboronic acid
Formula: C₈H₈BNO₆
Peso molecolare: 224,96 g/mol
Punto di fusione: 168°C
MDL Number: MFCD01632202
CAS Number: 85107-55-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR