Methyl-4-oxopiperidine-3-carboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 4-oxopiperidine-3-carboxylate hydrochloride, Methyl 4-Oxopiperidine-3-carboxylate HCl
M2852-5G
M2852-25G
TCIAM2852-5GEA
86
EUR
TCIAM2852-5G
TCIAM2852-25G
Methyl-4-oxopiperidine-3-carboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Methyl-4-oxopiperidine-3-carboxylate hydrochloride
Formula:
C₇H₁₂ClNO₃ Peso molecolare: 193,63 g/mol |
MDL Number:
MFCD00012769 CAS Number: 71486-53-8 EINECS: 275-526-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...