Stampa…

2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4-ol, 4-Hydroxy-2-(methylthio)-6-(trifluoromethyl)pyrimidine, 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one

M2851-1G M2851-5G
TCIAM2851-1GEA 23 EUR
TCIAM2851-1G TCIAM2851-5G
2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinol ≥98.0% (by GC)
2-(Methylsulphanyl)-6-(trifluoromethyl)-4-pyrimidinol
Formula: C₆H₅F₃N₂OS
Peso molecolare: 210,18 g/mol
MDL Number: MFCD00153191
CAS Number: 16097-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d21a1629fc3de3adc8cf633f75fde8e4e9c2ba96&vid=d21a1629fc3de3adc8cf633f75fde8e4e9c2ba96&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 177.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR