Stampa…

4-Methoxy-N-methylbenzylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

M2836-1G M2836-5G
TCIAM2836-1GEA 29 EUR
TCIAM2836-1G TCIAM2836-5G
4-Methoxy-N-methylbenzylamine ≥98.0% (by GC, titration analysis)
4-Methoxy-N-methylbenzylamine
Formula: C₉H₁₃NO
Peso molecolare: 151,21 g/mol
Punto di ebollizione: 238°C
MDL Number: MFCD04115407
CAS Number: 702-24-9
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c493af386cd3bf45be37ef73751197f340462a2b&vid=c493af386cd3bf45be37ef73751197f340462a2b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR