Stampa…

Methyl-4-aminopicolinate ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-Aminopyridine-2-carboxylic acid methyll ester, Methyl 4-Aminopyridine-2-carboxylate, 4-Aminopicolinic acid methyll ester, Methyl 4-Aminopicolinate

M2833-1G M2833-5G
TCIAM2833-1GEA 38 EUR
TCIAM2833-1G TCIAM2833-5G
Methyl-4-aminopicolinate ≥98.0% (by GC, titration analysis)
Methyl-4-aminopicolinate
Formula: C₇H₈N₂O₂
Peso molecolare: 152.1500 g/mol
MDL Number: MFCD00956000
CAS Number: 71469-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=136de88c658c6379bc9f993a30bf5fa72ce4cab5&vid=136de88c658c6379bc9f993a30bf5fa72ce4cab5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR