Methyl-1H-indazole-6-carboxylate ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl 1H-indazole-6-carboxylate, Indazole-6-carboxylic acid methyll ester, Methyl Indazole-6-carboxylate
M2823-1G
M2823-5G
TCIAM2823-1GEA
66
EUR
TCIAM2823-1G
TCIAM2823-5G
Methyl-1H-indazole-6-carboxylate ≥97.0% (by GC)
Methyl-1H-indazole-6-carboxylate
Formula:
C₉H₈N₂O₂ Peso molecolare: 176,17 g/mol |
MDL Number:
MFCD07371612 CAS Number: 170487-40-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...