Stampa…

2-Methoxycarbazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

M2817-1G M2817-5G
TCIAM2817-1GEA 76 EUR
TCIAM2817-1G TCIAM2817-5G
2-Methoxycarbazole ≥98.0% (by GC)
2-Methoxycarbazole
Formula: C₁₃H₁₁NO
MDL Number: MFCD09033506
CAS Number: 6933-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3df288bab177e1885b178d2dff3a633d2316b404&vid=3df288bab177e1885b178d2dff3a633d2316b404&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 237.0 to 241.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR