Stampa…

5-Methoxypyridine-3-boronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

M2810-1G M2810-5G
TCIAM2810-1GEA 93 EUR
TCIAM2810-1G TCIAM2810-5G
5-Methoxypyridine-3-boronic acid (contains varying amounts of Anhydride)
5-Methoxypyridine-3-boronic acid
Formula: C₆H₈BNO₃
Peso molecolare: 152,95 g/mol
MDL Number: MFCD03425977
CAS Number: 850991-69-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1d305273c692518a807b821a39832f90927f87b&vid=d1d305273c692518a807b821a39832f90927f87b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR