Stampa…

3-Methoxycarbonyl-5-nitrobenzeneboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, 3-Methoxycarbonyl-5-nitrophenylboronic acid, 3-(Methoxycarbonyl)-5-nitrophenylboronic acid, (3-(Methoxycarbonyl)-5-nitrophenyl)boronic acid

M2808-1G M2808-5G
TCIAM2808-1GEA 62 EUR
TCIAM2808-1G TCIAM2808-5G
3-Methoxycarbonyl-5-nitrobenzeneboronic acid (contains varying amounts of Anhydride)
3-Methoxycarbonyl-5-nitrobenzeneboronic acid
Formula: C₈H₈BNO₆
Peso molecolare: 224,96 g/mol
Punto di fusione: 224°C
MDL Number: MFCD02179469
CAS Number: 117342-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=626e028585d095e520823f59e7e8f69d7f104e2f&vid=626e028585d095e520823f59e7e8f69d7f104e2f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 95.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR