Methyl-3,4-dimethoxybenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl veratrate, 3,4-Dimethoxybenzoic acid methyl ester, Methyl 3,4-dimethoxybenzoate
M2805-5G
M2805-25G
TCIAM2805-5GEA
19
EUR
TCIAM2805-5G
TCIAM2805-25G
Methyl-3,4-dimethoxybenzoate ≥98.0% (by GC)
Methyl-3,4-dimethoxybenzoate
Formula:
C₁₀H₁₂O₄ Peso molecolare: 196,2 g/mol Punto di ebollizione: 283°C |
MDL Number:
MFCD00008430 CAS Number: 2150-38-1 EINECS: 218-424-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...