Stampa…

2-Methyl-5-nitrobenzotrifluoride ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-Methyl-4-nitro-2-(trifluoromethyl)benzene

M2798-1G M2798-5G
TCIAM2798-1GEA 21 EUR
TCIAM2798-1G TCIAM2798-5G
2-Methyl-5-nitrobenzotrifluoride ≥98.0% (by GC)
2-Methyl-5-nitrobenzotrifluoride
Formula: C₈H₆F₃NO₂
Peso molecolare: 205,14 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01631684
CAS Number: 89976-12-5
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR