Stampa…

Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-(Boc-amino)-3-nitrobenzoic acid methyl ester, 2-(tert-Butoxycarbonylamino)-3-nitrobenzoic acid methyl ester, Methyl 2-(Boc-amino)-3-nitrobenzoate, Methyl 2-((tert-butoxycarbonyl)amino)-3-nitrobenzoate

M2797-5G M2797-25G
TCIAM2797-5GEA 86 EUR
TCIAM2797-5G TCIAM2797-25G
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate ≥98.0% (by GC)
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate
Formula: C₁₃H₁₆N₂O₆
Peso molecolare: 296,28 g/mol
MDL Number: MFCD06797690
CAS Number: 57113-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f62795d274abb8f0fb732b408cfa7b442933c5b&vid=0f62795d274abb8f0fb732b408cfa7b442933c5b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR