Stampa…

Methyl-2-amino-5-hydroxybenzoate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Hydroxyanthranilic acid methyll ester, Methyl 5-Hydroxyanthranilate, Methyl 2-amino-5-hydroxybenzoate, Methyl 2-Amino-5-hydroxybenzoate, 2-Amino-5-hydroxybenzoic acid methyll ester

M2796-1G M2796-5G
TCIAM2796-1GEA 42 EUR
TCIAM2796-1G TCIAM2796-5G
Methyl-2-amino-5-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Methyl-2-amino-5-hydroxybenzoate
Formula: C₈H₉NO₃
Peso molecolare: 167,1600 g/mol
MDL Number: MFCD05863263
CAS Number: 1882-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8358d1688fb68f579f99c9b00ed032b553aaca2b&vid=8358d1688fb68f579f99c9b00ed032b553aaca2b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR