Stampa…

Methyl 3,5-Dihydroxy-4-methoxybenzoate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

M2794-1G M2794-5G
TCIAM2794-1GEA 147 EUR
TCIAM2794-1G TCIAM2794-5G
Methyl 3,5-Dihydroxy-4-methoxybenzoate ≥98.0% (by GC)
Methyl 3,5-Dihydroxy-4-methoxybenzoate
Formula: C₉H₁₀O₅
MDL Number: MFCD00017195
CAS Number: 24093-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01eca4f5f981d4dac9f789d535d51325bb6036dc&vid=01eca4f5f981d4dac9f789d535d51325bb6036dc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR