Stampa…

Methyl-3,3,3-trifluoropropanate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Methyl 3,3,3-Trifluoropropionate, 3,3,3-Trifluoropropionic acid methyll ester

M2783-1G M2783-5G
TCIAM2783-1GEA 60 EUR
TCIAM2783-1G TCIAM2783-5G
Methyl-3,3,3-trifluoropropanate ≥98.0% (by GC)
Methyl-3,3,3-trifluoropropanate
Formula: C₄H₅F₃O₂
Peso molecolare: 142,08 g/mol
Punto di ebollizione: 96°C
MDL Number: MFCD02262206
CAS Number: 18830-44-9
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6183f852aab534446b8ea7dc50fbae8b2315013a&vid=6183f852aab534446b8ea7dc50fbae8b2315013a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR