Stampa…

N-Methyl-N-[4-(trifluoromethyl)phenyl]amine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-Methyl-4-(trifluoromethyl)aniline

M2774-1G M2774-5G
TCIAM2774-1GEA 41 EUR
TCIAM2774-1G TCIAM2774-5G
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine ≥98.0% (by GC, titration analysis)
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine
Formula: C₈H₈F₃N
Peso molecolare: 175,15 g/mol
Punto di ebollizione: 106°C
Flash Pt: 98°C
MDL Number: MFCD07369717
CAS Number: 22864-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5470f9f1316b2437313b70b0727b56bfc382801&vid=e5470f9f1316b2437313b70b0727b56bfc382801&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR