Stampa…

1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Carbamoyl-1-methyl-4-nitro-3-propylpyrazole, 1-Methyl-4-nitro-3-propyl-(1H)-pyrazole-5-carboxamide

M2773-5G M2773-25G
TCIAM2773-5GEA 133 EUR
TCIAM2773-5G TCIAM2773-25G
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide ≥96.0% (by GC)
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
Formula: C₈H₁₂N₄O₃
Peso molecolare: 212,21 g/mol
MDL Number: MFCD02253196
CAS Number: 139756-01-7
EINECS: 423-960-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=256c5bbc870e2c4f2548319d6f48b4c3e1b41b11&vid=256c5bbc870e2c4f2548319d6f48b4c3e1b41b11&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 137.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR