Stampa…

Methyl-4-ethynylbenzoate ≥98.0% (by GC)

Fornitore: TCI

Synonyms: Methyl 4-ethynylbenzoate

M2768-1G M2768-5G
TCIAM2768-1GEA 83 EUR
TCIAM2768-1G TCIAM2768-5G
Methyl-4-ethynylbenzoate ≥98.0% (by GC)
Methyl-4-ethynylbenzoate
Formula: C₁₀H₈O₂
Peso molecolare: 160.17 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00168820
CAS Number: 3034-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561fd9c63616bf11b15af35389e449e4fd0e5f3c&vid=561fd9c63616bf11b15af35389e449e4fd0e5f3c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR