6-Methylflavone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
6-Methyl-2-phenylchromen-4-one, 6-Methyl-2-phenyl-4H-chromen-4-one, 6-Methyl-2-phenyl-4H-benzo[b]pyran-4-one, 6-Methyl-2-phenyl-4H-1-benzopyran-4-one, 6-Methyl-2-phenyl-4-benzopyrone
M2766-1G
M2766-5G
TCIAM2766-1GEA
44
EUR
TCIAM2766-1G
TCIAM2766-5G
6-Methylflavone ≥98.0% (by GC)
6-Methylflavone
Formula:
C₁₆H₁₂O₂ Peso molecolare: 236,27 g/mol Punto di ebollizione: 235°C |
MDL Number:
MFCD00017461 CAS Number: 29976-75-8 |
Risultati del test di specificazione
Appearance | White to Green to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 119.0 to 123.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...