6-Methylchromone ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
M2764-1G
M2764-5G
TCIAM2764-1GEA
30
EUR
TCIAM2764-1G
TCIAM2764-5G
6-Methylchromone ≥98.0% (by GC)
6-Methylchromone
Formula:
C₁₀H₈O₂ |
MDL Number:
MFCD00218598 CAS Number: 38445-23-7 |
|
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...