2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
M2760-1G
M2760-5G
TCIAM2760-1GEA
87
EUR
TCIAM2760-1G
TCIAM2760-5G
2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥97.0% (by HPLC, titration analysis)
2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Formula:
C₈H₇N₃O₂ |
MDL Number:
MFCD19443280 CAS Number: 739364-95-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...