Stampa…

Methyl-3-methylpicolinate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 3-Methylpicolinate, Methyl 3-Methylpyridine-2-carboxylate, Methyl 3-methylpicolinate, 3-Methylpyridine-2-carboxylic acid methyll ester, 3-Methylpicolinic acid methyll ester

M2746-1G M2746-5G
TCIAM2746-1GEA 26 EUR
TCIAM2746-1G TCIAM2746-5G
Methyl-3-methylpicolinate ≥98.0% (by GC)
Methyl-3-methylpicolinate
Formula: C₈H₉NO₂
Peso molecolare: 151,1600 g/mol
Punto di ebollizione: 73°C
MDL Number: MFCD03789583
CAS Number: 59718-84-2

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR