Stampa…

Methyl-6-hydroxynicotinate ≥97.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Methyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate, Methyl 6-hydroxynicotinate

M2727-1G M2727-5G
TCIAM2727-1GEA 53 EUR
TCIAM2727-1G TCIAM2727-5G
Methyl-6-hydroxynicotinate ≥97.0% (by GC)
Methyl-6-hydroxynicotinate
Formula: C₇H₇NO₃
MDL Number: MFCD04113616
CAS Number: 66171-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e266128323fe5fa44e1f7af440feb65bfb419ea&vid=4e266128323fe5fa44e1f7af440feb65bfb419ea&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR