Stampa…

6-Methylchromone-2-carboxylic acid ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

M2725-1G M2725-5G
TCIAM2725-1GEA 83 EUR
TCIAM2725-1G TCIAM2725-5G
6-Methylchromone-2-carboxylic acid ≥98.0% (by HPLC)
6-Methylchromone-2-carboxylic acid
Formula: C₁₁H₈O₄
Punto di fusione: 258°C
MDL Number: MFCD00239435
CAS Number: 5006-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8703e3470d0f104609da43f9a19bb8e7e9ad667&vid=d8703e3470d0f104609da43f9a19bb8e7e9ad667&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR