Stampa…

Methyl-3-hydroxy-2-thiophenecarboxylate ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 3-hydroxythiophene-2-carboxylate

M2716-1G M2716-5G
TCIAM2716-1GEA 27 EUR
TCIAM2716-1G TCIAM2716-5G
Methyl-3-hydroxy-2-thiophenecarboxylate ≥97.0% (by GC)
Methyl-3-hydroxy-2-thiophenecarboxylate
Formula: C₆H₆O₃S
Peso molecolare: 158,18 g/mol
Punto di ebollizione: 108°C
MDL Number: MFCD00055642
CAS Number: 5118-06-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5690569b5b443cf2d884defa8cd4e13e52ba116d&vid=5690569b5b443cf2d884defa8cd4e13e52ba116d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR