Stampa…

2-Methyl-4(1H)-quinazolinone ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M2709-5G M2709-25G
TCIAM2709-5GEA 85 EUR
TCIAM2709-5G TCIAM2709-25G
2-Methyl-4(1H)-quinazolinone ≥98.0% (by GC, titration analysis)
2-Methyl-4(1H)-quinazolinone
Formula: C₉H₈N₂O
MDL Number: MFCD01457186
CAS Number: 1769-24-0
EINECS: 217-189-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04b4a61e859a389e71d399cb33a108ca2675a227&vid=717ab02366b6f6f49a947da74d52b44daac9e7f6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 238.0 to 242.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR