Stampa…

4-Methyl-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

M2708-1G M2708-5G
TCIAM2708-1GEA 41 EUR
TCIAM2708-1G TCIAM2708-5G
4-Methyl-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
Peso molecolare: 204,15 g/mol
MDL Number: MFCD01631592
CAS Number: 261952-01-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7828861a04c776f34753cf7737e89a03b3a671a0&vid=7828861a04c776f34753cf7737e89a03b3a671a0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR