1-Methylisatin ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Methylindoline-2,3-dione, 1-Methyl-2,3-dihydroindole-2,3-dione, N-Methylindoline-2,3-dione, N-Methylisatin, 1-Methyl-1H-indole-2,3-dione, N-Methyl-1H-indole-2,3-dione, N-Methyl-2,3-dihydroindole-2,3-dione
M2706-1G
M2706-5G
TCIAM2706-1GEA
38
EUR
TCIAM2706-1G
TCIAM2706-5G
1-Methylisatin ≥98.0% (by GC)
1-Methylisatin
Formula:
C₉H₇NO₂ Peso molecolare: 161,16 g/mol |
MDL Number:
MFCD00005812 CAS Number: 2058-74-4 EINECS: 218-164-9 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 133.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...