Methylcyclopentadiene dimer (so called) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Stabilization:
stabilizzato con BHT/Ionolo
M2705-100ML
M2705-500ML
TCIAM2705-100MLEA
17
EUR
TCIAM2705-100ML
TCIAM2705-500ML
Methylcyclopentadiene dimer (so called) stabilizzato
Methylcyclopentadiene dimer
Formula:
C₁₂H₁₆ Peso molecolare: 160,26 g/mol Flash Pt: 53°C |
MDL Number:
MFCD00082297 CAS Number: 26472-00-4 EINECS: 247-724-5 UN: 3295 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...