Cloruro di metiltrifenilfosfonio ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Methyltriphenylphosphonium chloride, Methyl-TPP-Chloride, MTPP-Chloride
Formula:
C₁₉H₁₈ClP Peso molecolare: 312.78 g/mol |
MDL Number:
MFCD00797851 CAS Number: 1031-15-8 EINECS: 418-400-2 UN: 3077 ADR: 9 III |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 221.0 to 225.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...