Ethyl hydrogen malonate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid
Formula:
C₅H₈O₄ Peso molecolare: 132.115 g/mol Punto di ebollizione: 208°C Punto di fusione: –13°C Flash Pt: 113°C |
MDL Number:
MFCD00020490 CAS Number: 1071-46-1 EINECS: 213-992-7 |
|
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...