Methyl-3-iodo-4-methylbenzoate ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
Methyl 3-iodo-4-methylbenzoate
Formula:
C₉H₉IO₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00230583 CAS Number: 90347-66-3 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...