Stampa…

Methyl-2-bromothiazole-5-carboxylate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 2-bromothiazole-5-carboxylate

M2669-1G M2669-5G
TCIAM2669-1GEA 27 EUR
TCIAM2669-1G TCIAM2669-5G
Methyl-2-bromothiazole-5-carboxylate ≥95.0% (by GC)
Methyl-2-bromothiazole-5-carboxylate
Formula: C₅H₄BrNO₂S
Peso molecolare: 222,05 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD03788565
CAS Number: 54045-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f6178fd9fd8cb59a9ac58828105b2adb7dab6b4&vid=7f6178fd9fd8cb59a9ac58828105b2adb7dab6b4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR