Stampa…

4-Methyloxazole-5-carboxylic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

M2663-200MG M2663-1G
TCIAM2663-200MGEA 65 EUR
TCIAM2663-200MG TCIAM2663-1G
4-Methyloxazole-5-carboxylic acid ≥98.0% (by titrimetric analysis)
4-Methyloxazole-5-carboxylic acid
Formula: C₅H₅NO₃
Peso molecolare: 127,1000 g/mol
MDL Number: MFCD01571070
CAS Number: 2510-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f98a79351ec54ef815feafd8b3a2622c2b8d322&vid=5f98a79351ec54ef815feafd8b3a2622c2b8d322&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR