S-(2-Methyl-3-furyl) Thioacetate ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methyl-3-furanthiol Acetate, Thioacetic acid S-(2-Methyl-3-furyl) ester, S-(2-Methylfuran-3-yl) ethanethioate, 3-Acetylthio-2-methylfuran
M2661-5G
M2661-25G
TCIAM2661-5GEA
25
EUR
TCIAM2661-5G
TCIAM2661-25G
S-(2-Methyl-3-furyl) Thioacetate ≥96.0% (by GC)
S-(2-Methyl-3-furyl) Thioacetate
Formula:
C₇H₈O₂S Peso molecolare: 156,20 g/mol Punto di ebollizione: 235°C Flash Pt: 60°C |
MDL Number:
MFCD01632595 CAS Number: 55764-25-5 EINECS: 259-801-0 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...